Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

198765

Sigma-Aldrich

2-(4-Bromophenoxy)ethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCH2CH2OH
CAS Number:
Molecular Weight:
217.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

bp

184 °C/20 mmHg (lit.)

mp

53-55 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

OCCOc1ccc(Br)cc1

InChI

1S/C8H9BrO2/c9-7-1-3-8(4-2-7)11-6-5-10/h1-4,10H,5-6H2

InChI key

QYIOGYCRGNHDNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

2-(4-Bromophenoxy)ethanol was used in the synthesis of lithium 2-(4-lithiophenoxy)ethoxide[1].

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of lithium ω-(m-and p-lithiophenoxy) alkoxides modified with magnesium 2-ethoxyethoxide. Crystal structures of bis [4-(2-hydroxyethoxy) phenyl] mercury and bis [4-(3-hydroxypropoxy) phenyl] mercury.
Journal of Organometallic Chemistry, 590(1), 63-70 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service