Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19823

Sigma-Aldrich

4-tert-Butylcyclohexano-15-crown-5

≥95.0% (GC), yellow

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O5
CAS Number:
Molecular Weight:
330.46
Beilstein/REAXYS Number:
1684723
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥95.0% (GC)

color

yellow

density

1.08 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)C1CCC2OCCOCCOCCOCCOC2C1

InChI

1S/C18H34O5/c1-18(2,3)15-4-5-16-17(14-15)23-13-11-21-9-7-19-6-8-20-10-12-22-16/h15-17H,4-14H2,1-3H3

InChI key

KEDVODGFVKTPLB-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service