Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197882

Sigma-Aldrich

1,3,3-Trimethoxybutane

97%

Synonym(s):

4-Methoxy-2-butanone dimethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$249.00

$249.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$249.00

About This Item

Linear Formula:
CH3C(OCH3)2CH2CH2OCH3
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$249.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

assay

97%

form

liquid

refractive index

n20/D 1.41 (lit.)

bp

61-63 °C/20 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

functional group

ether
ketal

SMILES string

COCCC(C)(OC)OC

InChI

1S/C7H16O3/c1-7(9-3,10-4)5-6-8-2/h5-6H2,1-4H3

InChI key

DIQSNTFKLAYVOT-UHFFFAOYSA-N

Application

1,3,3-Trimethoxybutane may be used in chemical synthesis studies.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service