About This Item
assay
95%
form
liquid
bp
125 °C/0.03 mmHg (lit.)
mp
48-55 °C (lit.)
SMILES string
Clc1cc(Cl)cc(Oc2cccc(C=O)c2)c1
InChI
1S/C13H8Cl2O2/c14-10-5-11(15)7-13(6-10)17-12-3-1-2-9(4-12)8-16/h1-8H
InChI key
BISWHYILBVQCRA-UHFFFAOYSA-N
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service