Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197513

Sigma-Aldrich

2,3-Cycloheptenopyridine

97%

Synonym(s):

6,7,8,9-Tetrahydro-5H-cyclohepta[b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N
CAS Number:
Molecular Weight:
147.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

refractive index

n20/D 1.54 (lit.)

bp

97-98 °C/11 mmHg (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

C1CCc2cccnc2CC1

InChI

1S/C10H13N/c1-2-5-9-6-4-8-11-10(9)7-3-1/h4,6,8H,1-3,5,7H2

InChI key

CULUYAUTCSKQNM-UHFFFAOYSA-N

Application

2,3-Cycloheptenopyridine may be used in chemical synthesis studies.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service