Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196983

Sigma-Aldrich

3,5-Dinitrobenzotrifluoride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CF3
CAS Number:
Molecular Weight:
236.10
Beilstein/REAXYS Number:
2058079
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

99%

form

solid

mp

47-51 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(cc(c1)C(F)(F)F)[N+]([O-])=O

InChI

1S/C7H3F3N2O4/c8-7(9,10)4-1-5(11(13)14)3-6(2-4)12(15)16/h1-3H

InChI key

QZADIXWDDVQVKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dinitrobenzotrifluoride forms Meisenheimer-type σ-complexes with various nucleophiles in solution and has been studied by nuclear magnetic resonance spectroscopy[1].

Application

3,5-Dinitrobenzotrifluoride was used in the synthesis of 4,4′-bis(3-amino-5-trifluoromethyl phenoxy)-biphenyl, an aromatic diamine monomer[2]. It was also used as internal standard during the determination of ethalfluralin and trifluralin in soil extracts by negative chemical ionization mass spectrometry[3].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meisenheimer complexes of some 1-substituted-3, 5-dinitrobenzenes.
Foreman MI and Foster R.
Canadian Journal of Chemistry, 47(5), 729-732 (1969)
SYNTHESIS AND CHARACTERIZATION OF ORGANO-SOLUBLE FLUORINATED POLYIMIDES FROM 4, 4'-BIS (3-AMINO-5-TRIFLUOROMETHYLPHENOXY)-BIPHENYL AND VARIOUS AROMATIC DIANHYDRIDES.
Wu G-I, et al.
Chinese Journal of Polymer Science, 23(01), 55-61 (2005)
Quantitative confirmation of ethalfluralin and trifluralin in soil extracts by negative chemical ionization mass spectrometry.
Moyer JR and Elder JL.
Journal of Agricultural and Food Chemistry, 32(4), 866-868 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service