Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

195103

Sigma-Aldrich

Mordant yellow 10

Dye content 85 %

Synonym(s):

5-(4-Sulfophenylazo)salicylic acid disodium salt, 5-(p-Sulfophenylazo)salicylic acid, disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N2Na2O6S
CAS Number:
Molecular Weight:
366.26
Colour Index Number:
14010
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 85%

SMILES string

[Na+].[Na+].Oc1ccc(cc1C([O-])=O)\N=N\c2ccc(cc2)S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Nigović et al.
Journal of pharmaceutical and biomedical analysis, 26(5-6), 987-994 (2001-10-16)
The electrochemical properties of a colon-targeted prodrug of 5-aminosalicylic acid (5-ASA), 2-hydroxy-5-[(4-sulfophenyl)azo]benzoic acid (SPSA), were investigated in aqueous solutions at glassy carbon electrodes using cyclic voltammetry and controlled potential electrolysis. The influence of the pH and experimental time domain on
R Brindha et al.
Chemosphere, 201, 77-95 (2018-03-09)
Inspired by the efficiency of the photo-Fenton process on oxidation of organic pollutants, we herein present the feasibility of visible light driven photo-Fenton process as a post treatment of biological method for the effective degradation and detoxification of monoazo dye

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service