Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194522

Sigma-Aldrich

2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate

95%

Synonym(s):

DMPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(SH)CH2SO3Na · H2O
CAS Number:
Molecular Weight:
228.29
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

assay

95%

form

solid

mp

229 °C (dec.) (lit.)

SMILES string

[Na+].[H]O[H].[O-]S(=O)(=O)CC(S)CS

InChI

1S/C3H8O3S3.Na.H2O/c4-9(5,6)2-3(8)1-7;;/h3,7-8H,1-2H2,(H,4,5,6);;1H2/q;+1;/p-1

InChI key

XMUHNMQFDVIWGU-UHFFFAOYSA-M

Application

Efficient chelating agent for heavy metals.

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holger A Scheidt et al.
Biophysical journal, 109(3), 586-594 (2015-08-06)
Many proteins are anchored to lipid bilayer membranes through a combination of hydrophobic and electrostatic interactions. In the case of the membrane-bound nonreceptor tyrosine kinase Src from Rous sarcoma virus, these interactions are mediated by an N-terminal myristoyl chain and
Christy C Bridges et al.
Toxicology and applied pharmacology, 285(2), 110-117 (2015-04-15)
Secretion of inorganic mercury (Hg(2+)) from proximal tubular cells into the tubular lumen has been shown to involve the multidrug resistance-associated protein 2 (Mrp2). Considering similarities in localization and substrate specificity between Mrp2 and the breast cancer resistance protein (Bcrp)
Mafumi Hishida et al.
Chemistry and physics of lipids, 188, 61-67 (2015-05-11)
Phase behavior and structural properties were examined for phospholipid bilayers having different headgroups (DMPC, DMPS and DMPE) with added n-alkanes to study effect of flexible additives. Change in the temperatures of main transition of the lipid/alkane mixtures against the length
Sardar E Gasanov et al.
PloS one, 10(6), e0129248-e0129248 (2015-06-20)
Cobra venom cytotoxins are basic three-fingered, amphipathic, non-enzymatic proteins that constitute a major fraction of cobra venom. While cytotoxins cause mitochondrial dysfunction in different cell types, the mechanisms by which cytotoxins bind to mitochondria remain unknown. We analyzed the abilities
Shuping Wang et al.
Biochimie, 106, 167-174 (2014-09-10)
Dimercaptopropanesulfonic acid (DMPS) has been approved for the treatment of arsenic poisoning through promoting arsenic excretion and modulating arsenic species. To clarify how DMPS regulates the excretion of arsenic species, we investigated the effects of DMPS on the biomethylation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service