Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191965

Sigma-Aldrich

Poly(1-vinylnaphthalene)

average Mn ~100,000, powder

Synonym(s):

Isotactic poly(α-vinylnaphthalene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C10H7)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mn ~100,000

refractive index

n20/D 1.6818

transition temp

Tg 145 °C

SMILES string

C=Cc1cccc2ccccc12

Looking for similar products? Visit Product Comparison Guide

Application

Used to measure optical Kerr effect. Also used as an efficient hole transporter.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies of the antenna effect in polymer molecules. 8. Photophysics of water-soluble copolymers of 1-naphthylmethyl methacrylate and acrylic acid
Guillet, J. E., & Rendall, W. A
Macromolecules, 19(1), 224-230 (1986)
Photophysics of films of poly (2-vinylnaphthalene) doped with pyrene and TCNB
Kim, N., & Webber, S. E.
Macromolecules, 15(2), 430-435 (1982)
Excimer Formation in Solutions of Vinylaromatic Polymers
Odani, H.
Bull. Inst. Chem. Res., Kyoto Univ., 51(6), 351-372 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service