Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

190292

Sigma-Aldrich

3-(Trifluoromethyl)benzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2OH
CAS Number:
Molecular Weight:
176.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

68 °C/2 mmHg (lit.)

density

1.295 g/mL at 25 °C (lit.)

SMILES string

OCc1cccc(c1)C(F)(F)F

InChI

1S/C8H7F3O/c9-8(10,11)7-3-1-2-6(4-7)5-12/h1-4,12H,5H2

InChI key

BXEHKCUWIODEDE-UHFFFAOYSA-N

General description

3-(Trifluoromethyl)benzyl alcohol enhances the charging of myoglobin noncovalent complex during electrospray ionization mass spectroscopy[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

183.2 °F - closed cup

flash_point_c

84 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shirley H Lomeli et al.
Journal of the American Society for Mass Spectrometry, 21(1), 127-131 (2009-10-27)
The addition of m-nitrobenzyl alcohol (m-NBA) was shown previously (Lomeli et al., J. Am. Soc. Mass Spectrom. 2009, 20, 593-596) to enhance multiple charging of native proteins and noncovalent protein complexes in electrospray ionization (ESI) mass spectra. Additional new reagents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service