Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

188883

Sigma-Aldrich

4,4′-DDOH

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)2CHCH2OH
CAS Number:
Molecular Weight:
267.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

100-102 °C (lit.)

SMILES string

OCC(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H12Cl2O/c15-12-5-1-10(2-6-12)14(9-17)11-3-7-13(16)8-4-11/h1-8,14,17H,9H2

InChI key

ZVIDYKRNLNAXFT-UHFFFAOYSA-N

General description

DDOH is a metabolite of DDT and forms conjugates with palmitic acid in DDT-treated female rats[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Eriksson et al.
Brain research, 514(1), 141-142 (1990-04-23)
DDT, 1,1,1-trichloro-2,2-bis(p-chlorophenyl)-ethane is one of the best-known insecticides which produces neurotoxic syndrome in mammals. DDT is also a world-wide environmental contaminant which is still used in several countries. We have previously reported on pronounced retention of DDT in the neonatal
Edward J Scollon et al.
Environmental toxicology and chemistry, 31(2), 336-346 (2011-11-03)
This study evaluated the interactions of flight, fasting, and 1,1,1-trichloro-bis(4-chlorophenyl)ethane (p,p'-DDT) loading on residue metabolism and distribution in recently exposed white-crowned sparrows (Zonotrichia leucophrys). Female sparrows were dosed with 5 mg p,p'-DDT per kg body weight over 3 d. Following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service