All Photos(1)
About This Item
Linear Formula:
O2NC6H4(CH2)4OH
CAS Number:
Molecular Weight:
195.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
form
solid
refractive index
n20/D 1.557 (lit.)
density
1.154 g/mL at 25 °C (lit.)
SMILES string
OCCCCc1ccc(cc1)[N+]([O-])=O
InChI
1S/C10H13NO3/c12-8-2-1-3-9-4-6-10(7-5-9)11(13)14/h4-7,12H,1-3,8H2
InChI key
MFYJBAQONVFIFC-UHFFFAOYSA-N
Application
4-(4-Nitrophenyl)-1-butanol was used in chemoenzymatic synthesis of bacterial O-antigen from Salmonella serogroup E1, 3-O-(4-O-β-D-mannopyranosyl-α-L-rhamnopyranosyl)-α-D-galactose.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y Zhao et al.
Carbohydrate research, 319(1-4), 184-191 (1999-10-16)
The chemical synthesis of the bacterial O-antigen from Salmonella serogroup E1, 3-O-(4-O-beta-D-mannopyranosyl-alpha-L-rhamnopyranosyl)-alpha-D-galactos e, presents a particular challenge because it contains a beta-(1-->4) mannosidic linkage to L-rhamnose. We report a chemoenzymatic synthesis of this crucial antigenic material which culminates in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service