Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187666

Sigma-Aldrich

(4S,5S)-(−)-2-Methyl-5-phenyl-2-oxazoline-4-methanol

98%

Synonym(s):

(4S,5S)-(−)-4-Hydroxymethyl-2-methyl-5-phenyloxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
Beilstein/REAXYS Number:
1212843
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

optical activity

[α]20/D −172°, c = 10.8 in chloroform

mp

61-65 °C (lit.)

SMILES string

CC1=N[C@@H](CO)[C@@H](O1)c2ccccc2

InChI

1S/C11H13NO2/c1-8-12-10(7-13)11(14-8)9-5-3-2-4-6-9/h2-6,10-11,13H,7H2,1H3/t10-,11-/m0/s1

InChI key

NTCJMVHDZUBYNA-QWRGUYRKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the asymmetric reduction of ketones to secondary alcohols.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service