Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187259

Sigma-Aldrich

3,4-Dibenzyloxybenzyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2O)2C6H3CH2Cl
CAS Number:
Molecular Weight:
338.83
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

41-43 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCc1ccc(OCc2ccccc2)c(OCc3ccccc3)c1

InChI

1S/C21H19ClO2/c22-14-19-11-12-20(23-15-17-7-3-1-4-8-17)21(13-19)24-16-18-9-5-2-6-10-18/h1-13H,14-16H2

InChI key

DDAGFZBQVSGXJN-UHFFFAOYSA-N

Application

3,4-Dibenzyloxybenzyl chloride was used to prepare benzylnitrile precursor, required for the synthesis of amidines[1].

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New non peptidic C5a receptor antagonists.
Astles PC, et al.
Bioorganic & Medicinal Chemistry Letters, 7(7), 907-912 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service