185973
Acetone-d6
99.5 atom % D
Synonym(s):
Hexadeuteroacetone
About This Item
Recommended Products
vapor density
2 (vs air)
vapor pressure
14.39 psi ( 55 °C)
15.07 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)
4.08 psi ( 20 °C)
isotopic purity
99.5 atom % D
assay
≥99.5%
contains
1% TMS
expl. lim.
13.2 %
packaging
ampule of 10 g (prescored)
bottle of 25 g, 50 g (screw cap)
refractive index
n20/D 1.355 (lit.)
bp
55.5 °C (lit.)
mp
−93.8 °C (lit.)
density
0.872 g/mL at 25 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
InChI
1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3
Inchi Key
CSCPPACGZOOCGX-WFGJKAKNSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service