Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is
Microwave spectrum and molecular structure of formamide.
Costain CC and Dowling JM.
J. Chem. Phys. , 32(1), 158-165 (1960)
The formamide method for the extraction of polysaccharides from haemolytic streptococci.
Fuller AT.
British Journal of Experimental Pathology, 32(1), 130-130 (1960)
Solubilization in formamide protects RNA from degradation.
Chomczynski P.
Nucleic Acids Research, 20(14), 3791-3791 (1992)
Formamide can dramatically improve the specificity of PCR.