Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

182540

Sigma-Aldrich

Poly(4-vinylbiphenyl)

average Mw ~115,000 by GPC, powder

Synonym(s):

4-Vinylbiphenyl homopolymer, Poly(4-phenylstyrene), Poly(4-vinylbiphenyl)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~115,000 by GPC

transition temp

Tg 138 °C

SMILES string

C=Cc1ccc(cc1)-c2ccccc2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen Rankin Willsey et al.
Neuron, 109(5), 788-804 (2021-01-27)
Gene Ontology analyses of autism spectrum disorders (ASD) risk genes have repeatedly highlighted synaptic function and transcriptional regulation as key points of convergence. However, these analyses rely on incomplete knowledge of gene function across brain development. Here we leverage Xenopus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service