Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18102

Sigma-Aldrich

Bromopolystyrene

extent of labeling: ~5 mmol/g Br loading

Synonym(s):

Copolymer of styrene and divinylbenzene, brominated, Polystyrene crosslinked with divinylbenzene, brominated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4Br)]n
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~5 mmol/g Br loading
90-95% substitution (brominated rings)

matrix

crosslinked with 2% DVB

particle size

100-200 mesh

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service