Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

180580

Sigma-Aldrich

Tricyclohexylmethanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3COH
CAS Number:
Molecular Weight:
278.47
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

94-96 °C (lit.)

functional group

hydroxyl

SMILES string

OC(C1CCCCC1)(C2CCCCC2)C3CCCCC3

InChI

1S/C19H34O/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h16-18,20H,1-15H2

InChI key

MRGVKQSSSOCONZ-UHFFFAOYSA-N

General description

The heat capacity of liquid-quenched glass of tricyclohexylmethanol was measured by adiabatic calorimetry[1].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhisa Yamamura et al.
The journal of physical chemistry. B, 116(13), 3938-3943 (2012-03-15)
Heat capacities of liquids and liquid-quenched glasses (LQGs) of dicyclorohexylmethanol (DCHM) and tricyclohexylmethanol (TCHM) were measured by adiabatic calorimetry. Upon cooling the liquid compounds, they undergo glass transitions around 250 and 265 K, respectively. Temperature dependence of the FT-IR spectrum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service