Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17984

Sigma-Aldrich

Z-D-β-Dab(Fmoc)-OH

purum, ≥98.0% (HPLC)

Synonym(s):

β-Z-γ-Fmoc-D-3,4-diaminobutyric acid, γ-Fmoc-β-Z-D-3,4-diaminobutyric acid, (S)-4-(Fmoc-amino)-3-(Z-amino)butyric acid, Z-D-β-Dbu(Fmoc)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26N2O6
CAS Number:
Molecular Weight:
474.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

grade

purum

assay

≥98.0% (HPLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

179-183 °C

application(s)

peptide synthesis

functional group

Fmoc

SMILES string

OC(=O)C[C@@H](CNC(=O)OCC1c2ccccc2-c3ccccc13)NC(=O)OCc4ccccc4

InChI

1S/C27H26N2O6/c30-25(31)14-19(29-27(33)34-16-18-8-2-1-3-9-18)15-28-26(32)35-17-24-22-12-6-4-10-20(22)21-11-5-7-13-23(21)24/h1-13,19,24H,14-17H2,(H,28,32)(H,29,33)(H,30,31)/t19-/m0/s1

Inchi Key

GNBVOVLCIBZKIQ-IBGZPJMESA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service