Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17894

Sigma-Aldrich

Polyethylene glycol monomethyl ether mesylate

2,000

Synonym(s):

mono-Methyl polyethylene glycol mesylate, O-Mesyl-O′-methyl-polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nSO2CH3
CAS Number:
MDL number:
UNSPSC Code:
12162002

mp

52-55 °C

Ω-end

mesylate

α-end

methoxy

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polymer support for PEG-supported synthesis

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X. Zhao et al.
Tetrahedron Letters, 38, 5437-5437 (1997)
Organic Synthesis on Soluble Polymer Supports: Liquid-Phase Methodologies.
Dennis J. Gravert et al.
Chemical reviews, 97(2), 489-510 (1997-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service