Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

176400

Sigma-Aldrich

Tetrafluoroboric acid dimethyl ether complex (1:1 mole complex)

Synonym(s):

(Dimethyl ether)hydrogen tetrafluoroborate, Dimethyloxonium tetrafluoroborate, Fluoroboric acid dimethyl ether complex, Tetrafluoroboric acid dimethyl etherate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HBF4 · O(CH3)2
CAS Number:
Molecular Weight:
133.88
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

density

1.1 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

F.COC.FB(F)F

InChI

1S/C2H6O.BF3.FH/c1-3-2;2-1(3)4;/h1-2H3;;1H

InChI key

ROVHHTLFIJTTKB-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-41.8 °F - closed cup

flash_point_c

-41 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service