Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17359

Sigma-Aldrich

O-[4-(Hydroxymethyl)benzoyl]-O′-methyl-polyethylene glycol 5000

≥80% (NMR)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

assay

≥80% (NMR)

mol wt

average Mw 5,000

mp

56-59 °C

Ω-end

4-(hydroxymethyl)benzoyl

α-end

methoxy

storage temp.

−20°C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayi Wu et al.
Journal of hepatology, 70(5), 918-929 (2018-12-26)
Liver cancer is the second leading cause of cancer death worldwide. Hepatocellular carcinoma (HCC) is the most common type of primary liver cancer in adults. The aim of this study was to define the role of the long non-coding RNA
Pingping Zhu et al.
Nature communications, 7, 13608-13608 (2016-12-03)
Liver cancer stem cells (CSCs) may contribute to the high rate of recurrence and heterogeneity of hepatocellular carcinoma (HCC). However, the biology of hepatic CSCs remains largely undefined. Through analysis of transcriptome microarray data, we identify a long noncoding RNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service