17337
Cyanoborohydride on polymer support
extent of labeling: 2.5-4.5 mmol/g BH3CN- loading
Synonym(s):
Amberlite™ IRA-900 BH3CN form
About This Item
Recommended Products
form
beads
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: imine reactive
extent of labeling
2.5-4.5 mmol/g BH3CN- loading
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service