About This Item
Recommended Products
assay
98%
reaction suitability
reaction type: C-C Bond Formation
mp
85-88 °C (lit.)
SMILES string
[Br-].CCCCCCCCCCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
InChI
1S/C30H40P.BrH/c1-2-3-4-5-6-7-8-9-10-20-27-31(28-21-14-11-15-22-28,29-23-16-12-17-24-29)30-25-18-13-19-26-30;/h11-19,21-26H,2-10,20,27H2,1H3;1H/q+1;/p-1
InChI key
NSIFOGPAKNSGNW-UHFFFAOYSA-M
Application
Intercalating agent for modifying anticorrosion, gas barrier, mechanical and thermal properties of nanocomposite coatings
Mediator for reactions of nitrophenyl acetate
Studies of:
- Nucleophilic substitution kinetics of sulfonate ester
- Intercalated polyester / silicate nanocomposites
- Thermal stability of quaternary phosphonium-modified montmorillonites in silicate layered polymer nanocomposites
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service