17098
Acetate on polymer support
extent of labeling: ~1.5 mmol/g CH3COO- loading
Synonym(s):
Ambersep® 900 acetate-form
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
PubChem Substance ID:
extent of labeling
~1.5 mmol/g CH3COO- loading
impurities
~20% water
anion traces
chloride (Cl-): ≤1000 mg/kg
Legal Information
Ambersep is a registered trademark of DuPont de Nemours, Inc.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service