Recommended Products
technique(s)
LPLC: suitable
size
5 cm × 10 cm
matrix active group
Silica
General description
FluoroFlash® TLC Plates uses fluorous silica as the stationary phase i.e., silica gel with a fluorocarbon bonded phase (-SiMe2(CH2)2C8F17). It is evident that more fluorinated products have smaller retention factor due to its strong interaction with fluorous plate.[1]
Application
FluoroFlash® TLC Plates were used in TLC analysis.[2]
Packaging
package with 10 pieces
Legal Information
Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents US 6,156,896; US 5,859,247; US 5,777,121 and US 6,673,539.
FluoroFlash is a registered trademark of Fluorous Technologies, Inc.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kenichi Hatanaka
Topics in current chemistry, 308, 291-306 (2011-10-06)
A series of fluorous-tagged glycosides with different number of fluorine atoms are incorporated into the cells, transported to Golgi, elongated by cellular enzymes, and then released to the culture medium. Fluorine content strongly affects on the affinity for cell membrane
H-J Lehmler et al.
Tetrahedron, 66(14), 2561-2569 (2010-03-23)
A series of colored hydrocarbon and fluorocarbon tagged 1-fluoro-4-alkylamino-anthraquinones and 1,4-bis-alkylamino-anthraquinone probe molecules were synthesized from a (fluorinated) alkyl amine and 1,4-difluoroanthraquinone to aid in the development of fluorous separation applications. The anthraquinones displayed stacking of the anthraquinone tricycle and
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service