Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16521

Sigma-Aldrich

tert-Butyl 2-isocyano-3-phenylpropionate

≥96.0% (GC)

Synonym(s):

DL-Phenylalanine tert-butyl ester isocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NC)COOC(CH3)3
CAS Number:
Molecular Weight:
231.29
Beilstein/REAXYS Number:
3971005
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥96.0% (GC)

refractive index

n20/D 1.493

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)C(Cc1ccccc1)[N+]#[C-]

InChI

1S/C14H17NO2/c1-14(2,3)17-13(16)12(15-4)10-11-8-6-5-7-9-11/h5-9,12H,10H2,1-3H3

InChI key

ZAWPEVUURWHHNZ-UHFFFAOYSA-N

Application

tert-Butyl 2-isocyano-3-phenylpropionate may be used in the synthesis of cyclic amino acids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

discolors on storage to deep yellow or deep brownish-yellow

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service