Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16474

Sigma-Aldrich

7-Bromobicyclo[2.2.1]heptane

technical, ≥90%

Synonym(s):

7-Bromonorbornane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11Br
CAS Number:
Molecular Weight:
175.07
Beilstein/REAXYS Number:
1919875
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥90%

refractive index

n20/D 1.518

bp

75 °C/20 mmHg (lit.)

density

1.38 g/mL at 20 °C (lit.)

SMILES string

BrC1[C@H]2CC[C@@H]1CC2

InChI

1S/C7H11Br/c8-7-5-1-2-6(7)4-3-5/h5-7H,1-4H2/t5-,6+,7?

InChI key

WHGPQZJYJWIBGZ-MEKDEQNOSA-N

Application

7-Bromobicyclo[2.2.1]heptane(7-Bromonorbornane) was used in the synthesis of methyl norbornane-7-carboxylate.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Bis (semiquinone) s and Their Electrochemical and Electron Paramagnetic Resonance Spectral Characterization.
Shultz DA, et al.
The Journal of Organic Chemistry, 63(25), 9462-9469 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service