Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16429

Sigma-Aldrich

1-(Diacetoxyiodo)-4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)benzene

technical, ≥90%

Synonym(s):

1-(Diacetoxyiodo)-4-(1H,1H,2H,2H-perfluorodecyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14F17IO4
CAS Number:
Molecular Weight:
768.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical

assay

≥90%

mp

172-177 °C (D)

SMILES string

CC([O-])=O.CC(=O)O[I+]c1ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc1

InChI

1S/C18H11F17IO2.C2H4O2/c1-8(37)38-36-10-4-2-9(3-5-10)6-7-11(19,20)12(21,22)13(23,24)14(25,26)15(27,28)16(29,30)17(31,32)18(33,34)35;1-2(3)4/h2-5H,6-7H2,1H3;1H3,(H,3,4)/q+1;/p-1

Inchi Key

SGTQRUYUYBGLJM-UHFFFAOYSA-M

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service