Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163724

Sigma-Aldrich

BES

Synonym(s):

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, N,N-Bis(2-hydroxyethyl)taurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO5S
CAS Number:
Molecular Weight:
213.25
Beilstein/REAXYS Number:
1781572
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99% NaOH basis (dry basis, titration)

form

powder, crystals or chunks

useful pH range

6.4-7.8

pKa (25 °C)

7.1

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

OCCN(CCO)CCS(O)(=O)=O

InChI

1S/C6H15NO5S/c8-4-1-7(2-5-9)3-6-13(10,11)12/h8-9H,1-6H2,(H,10,11,12)

InChI key

AJTVSSFTXWNIRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer useful in the physiological pH range.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingjie Zhang et al.
ACS nano, 9(10), 10445-10452 (2015-09-25)
Artificial solids composed of semiconductor quantum dots (QDs) are being developed for large-area electronic and optoelectronic applications, but these materials often have defect-induced in-gap states (IGS) of unknown chemical origin. Here we performed scanning probe based spectroscopic analysis and density
Emma J Gagen et al.
FEMS microbiology letters, 362(14) (2015-06-26)
Molecular information suggests that there is a broad diversity of acetogens in the rumen, distinct from any currently isolated acetogens. We combined molecular analysis with enrichment culture techniques to investigate this diversity further. Methane-inhibited, hydrogenotrophic enrichment cultures produced acetate as
Yingwen Cheng et al.
Advanced materials (Deerfield Beach, Fla.), 27(42), 6598-6605 (2015-10-01)
An interface promoted approach is developed for guiding the design of stable and high capacity materials for Mg batteries using SnSb alloys as model materials. Experimental and theoretical studies reveal that the SnSb alloy has exceptionally high reversible capacity (420
R N Roy et al.
Cryobiology, 22(6), 589-600 (1985-12-01)
Information is given concerning two standard buffer solutions suitable as pH references in 30, 40, and 50 mass% dimethyl sulfoxide (DMSO)/H2O mixed solvents at subzero temperatures from -20 to 0 degrees C, with the intention of establishing a pH (designated
D L Freedman et al.
Applied and environmental microbiology, 55(9), 2144-2151 (1989-09-01)
A biological process for remediation of groundwater contaminated with tetrachloroethylene (PCE) and trichloroethylene (TCE) can only be applied if the transformation products are environmentally acceptable. Studies with enrichment cultures of PCE- and TCE-degrading microorganisms provide evidence that, under methanogenic conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service