Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

156531

Sigma-Aldrich

Diethyl (ethylthiomethyl)phosphonate

97%

Synonym(s):

NSC 140826

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
212.25
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.464 (lit.)

density

1.097 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CSCC)OCC

InChI

1S/C7H17O3PS/c1-4-9-11(8,10-5-2)7-12-6-3/h4-7H2,1-3H3

InChI key

FTTKKJQNGIJGOC-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of spiro and fused N,O-heterocyclic phosphonate esters as antibacterial and antifungal agents
  • Preparation of spiro-substituted pyrimidine phosphonate esters as antitumor agents
  • Stereoselective Horner-Wadsworth-Emmons reactions

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service