Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

156302

Sigma-Aldrich

Benzene

ReagentPlus®, thiophene free, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein/REAXYS Number:
969212
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

product line

ReagentPlus®

assay

≥99%

form

liquid

autoignition temp.

1043 °F

expl. lim.

8 %

refractive index

n20/D 1.501 (lit.)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

12.2 °F

flash_point_c

-11 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John C Thomas et al.
ACS nano, 9(5), 4734-4742 (2015-04-14)
Carboranethiol molecules self-assemble into upright molecular monolayers on Au{111} with aligned dipoles in two dimensions. The positions and offsets of each molecule's geometric apex and local dipole moment are measured and correlated with sub-Ångström precision. Juxtaposing simultaneously acquired images, we
Jelle Vlaanderen et al.
Environmental health perspectives, 119(2), 159-167 (2010-10-01)
The use of occupational cohort studies to assess the association of benzene and lymphoma is complicated by problems with exposure misclassification, outcome classification, and low statistical power. We performed meta-analyses of occupational cohort studies for five different lymphoma categories: Hodgkin
Cliona M McHale et al.
Carcinogenesis, 33(2), 240-252 (2011-12-15)
Benzene causes acute myeloid leukemia and probably other hematological malignancies. As benzene also causes hematotoxicity even in workers exposed to levels below the US permissible occupational exposure limit of 1 part per million, further assessment of the health risks associated
Audrey Smargiassi et al.
Environmental research, 132, 38-45 (2014-04-20)
The acute cardiorespiratory effects of air quality among children living in areas with considerable heavy industry have not been well investigated. We conducted a panel study of children with asthma living in proximity to an industrial complex housing two refineries
Subhamay Pramanik et al.
Analytica chimica acta, 793, 99-106 (2013-08-21)
Aggregates of hexaphenylbenzene derivatives 3, having pyrene groups form network of fluorescent nanofibres in presence of mercury ions. Further, fluorescent nanofibres of 3-Hg(2+) supramolecular ensemble exhibit sensitive and pronounced response towards the picric acid. In addition, the solution coated paper

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service