Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15534

Sigma-Aldrich

Boc-His(Bzl)-OH

≥98.0% (TLC)

Synonym(s):

Nα-Boc-N(im)-benzyl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23N3O4
CAS Number:
Molecular Weight:
345.39
Beilstein/REAXYS Number:
710413
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

assay

≥98.0% (TLC)

optical activity

[α]20/D +23±1°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

181-184 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1cn(Cc2ccccc2)cn1)C(O)=O

InChI

1S/C18H23N3O4/c1-18(2,3)25-17(24)20-15(16(22)23)9-14-11-21(12-19-14)10-13-7-5-4-6-8-13/h4-8,11-12,15H,9-10H2,1-3H3,(H,20,24)(H,22,23)/t15-/m0/s1

InChI key

OUHPNBGKEMHUCQ-HNNXBMFYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service