Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15433

Sigma-Aldrich

Boc-Thr(t-Bu)-OH

≥99.0% (sum of enantiomers, TLC)

Synonym(s):

Boc-O-tert-butyl-L-threonine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH(CH3)CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
275.34
Beilstein/REAXYS Number:
4454820
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0% (sum of enantiomers, TLC)

form

solid

optical activity

[α]20/D +3.6±0.5°, c = 2% in DMF

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C13H25NO5/c1-8(18-12(2,3)4)9(10(15)16)14-11(17)19-13(5,6)7/h8-9H,1-7H3,(H,14,17)(H,15,16)/t8-,9+/m1/s1

InChI key

LKRXXARJBFBMCE-BDAKNGLRSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Morales Torres et al.
Nature communications, 11(1), 1792-1792 (2020-04-15)
Continuous cancer growth is driven by subsets of self-renewing malignant cells. Targeting of uncontrolled self-renewal through inhibition of stem cell-related signaling pathways has proven challenging. Here, we show that cancer cells can be selectively deprived of self-renewal ability by interfering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service