Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15376

Sigma-Aldrich

Boc-Cys(Acm)-OH

≥96.0% (T)

Synonym(s):

Boc-S-acetamidomethyl-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2SCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
292.35
Beilstein/REAXYS Number:
2058303
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥96.0% (T)

optical activity

[α]20/D −36.5±1.5°, c = 1% in H2O

mp

111-114 °C

application(s)

peptide synthesis

SMILES string

CC(=O)NCSC[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C11H20N2O5S/c1-7(14)12-6-19-5-8(9(15)16)13-10(17)18-11(2,3)4/h8H,5-6H2,1-4H3,(H,12,14)(H,13,17)(H,15,16)/t8-/m0/s1

InChI key

HLCTYBOTPCIHTG-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marzieh Najafi et al.
Biomacromolecules, 19(9), 3766-3775 (2018-08-14)
In this study, native chemical ligation (NCL) was used as a selective cross-linking method to form core-cross-linked thermosensitive polymeric micelles for drug delivery applications. To this end, two complementary ABA triblock copolymers having polyethylene glycol (PEG) as midblock were synthesized
Wenjun Jiang et al.
Cell discovery, 3, 17037-17037 (2017-10-21)
The construction of mirror-image biological systems may open the next frontier for biomedical technology development and discovery. Here we have designed and chemically synthesized a mutant version of the thermostable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service