Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

153672

Sigma-Aldrich

Methyl 2-phenyl-4-quinolinecarboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13NO2
CAS Number:
Molecular Weight:
263.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

mp

60-62 °C (lit.)

SMILES string

COC(=O)c1cc(nc2ccccc12)-c3ccccc3

InChI

1S/C17H13NO2/c1-20-17(19)14-11-16(12-7-3-2-4-8-12)18-15-10-6-5-9-13(14)15/h2-11H,1H3

InChI key

FDNMMBMNYRTPLC-UHFFFAOYSA-N

Application

Methyl 2-phenyl-4-quinolinecarboxylate has been used to prepare 4-[1-(1H-Imidazol-4-yl)-ethyl]-2-phenyl-quinoline dihydrochloride.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia I Solomatina et al.
Dalton transactions (Cambridge, England : 2003), 49(20), 6751-6763 (2020-05-07)
A family of diimine (N^N) and cyclometalating (N^C) ligands based on a phenanthro-imidazole aromatic system: 2-pyridyl-1H-phenanthro[9,10-d]imidazole (N^N); 2-R-1-phenyl-1H-phenanthro[9,10-d]imidazole, R = phenyl (N^C4), 3-iodophenyl (N^C5) and 4-nitrophenyl (N^C6) were prepared. It was found that N^C4 and N^C5 show π-π* fluorescence typical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service