Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15307

Sigma-Aldrich

5-(Boc-amino)-1-pentanol

≥95.0% (GC)

Synonym(s):

tert-Butyl N-(5-hydroxypentyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNH(CH2)5OH
CAS Number:
Molecular Weight:
203.28
Beilstein/REAXYS Number:
3603659
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95.0% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.45

density

1.00 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCCCO

InChI

1S/C10H21NO3/c1-10(2,3)14-9(13)11-7-5-4-6-8-12/h12H,4-8H2,1-3H3,(H,11,13)

InChI key

DDGNGFVNTZJMMZ-UHFFFAOYSA-N

Application

5-(Boc-amino)-1-pentanol may also be used as non-limiting linkers.
5-(Boc-amino)-1-pentanol may be used as a precursor for the synthesis of hydroxylamino-containing 1-guanidine-7-aminopentane (GC7) analogs.[1]

Other Notes

Linker (anthraquinone-oligopeptide)[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Maruyama et al.
The Journal of Organic Chemistry, 57, 6143-6143 (1992)
Novel hydroxylamine-containing analogues of 1-guanidino-7-aminoheptane (GC7), an effective inhibitor of deoxyhypusine synthase.
Khomutov MA, et al.
Russian Journal of Bioorganic Chemistry, 42(4), 415-422 (2016)
Cheng-Ying Shen et al.
Experimental & applied acarology, 80(3), 381-398 (2020-02-08)
Sensitization to mites is a considerable factor in the development of allergic diseases. Because of its abundance, Tyrophagus putrescentiae (Tp) is the predominant storage mite found in home storage rooms, kitchens, and bakeries. Patients allergic to mites might exhibit a
Rosa Freitas et al.
Environmental pollution (Barking, Essex : 1987), 251, 756-765 (2019-05-24)
In the present study Mytilus galloprovincialis mussels were exposed for 28 days to three salinities: 30 (control), 25 and 35. Simultaneously, organisms at each salinity were exposed to either the antimicrobial agent Triclosan (TCS) or the pharmaceutical drug Diclofenac (DIC)
Yang Dong et al.
Current biology : CB, 29(6), 1038-1046 (2019-03-05)
Evolution of gene-regulatory sequences is considered the primary driver of morphological variation [1-3]. In animals, the diversity of body plans between distantly related phyla is due to the differential expression patterns of conserved "toolkit" genes [4]. In plants, variation in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service