Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152323

Sigma-Aldrich

Pyridine-d5

99 atom % D

Synonym(s):

Pentadeuteropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5N
CAS Number:
Molecular Weight:
84.13
Beilstein/REAXYS Number:
114377
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99 atom % D

assay

99% (CP)

expl. lim.

0.34-6.3 % (lit.)

impurities

≤0.05% water
water

refractive index

n20/D 1.506 (lit.)

bp

114.4 °C (lit.)

mp

-41 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D

Inchi Key

JUJWROOIHBZHMG-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine-d5 is a deuterated form of pyridine having an isotopic purity of 99atom%D. The reaction between pyridine vapor and heavy water in the presence of a palladium catalyst has been reported to afford pyridine-d5. Infrared (IR), Raman (in the range 300-4000cm-1) and high-resolution (0.06cm-1) FT-IR (Fourier transform infrared) spectral studies of pyridine-d5 have been reported.

Pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

62.6 °F

flash_point_c

17 °C

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectra of C2v deuterium substituted pyridines. 4-pyridine-2, 3, 5, 6-d4 and pyridine-d5.
Stidham HD and Dilella DP.
Journal of Raman Spectroscopy, 9(4), 247-256 (1980)
The FT-IR spectra of pyridine and pyridine-d51.
Wong KN and Colson SD.
Journal of Molecular Spectroscopy, 104(1), 129-151 (1984)
Xu Pang et al.
Carbohydrate research, 402, 236-240 (2014-12-17)
Five new steroidal saponins (1-5) were isolated from the fermentation broth of total furostanol glycosides from tubers of Dioscorea zingiberensis C.H. Wright incubated with a fungal, Absidia coerulea AS 3.3389, along with known saponins, zingiberensis new saponin (6), deltonin (7)
Xu Pang et al.
Steroids, 93, 68-76 (2014-12-03)
For the first time, a systematic phytochemical study was performed on the roots of Marsdenia tenacissima. Finally, sixteen new polyoxypregnane glycosides, marstenacissides A1-A7 (1-7) and marstenacissides B1-B9 (8-16), were isolated from M. tenacissima roots. The structures of these new compounds
Indra Prakash et al.
Natural product communications, 9(8), 1135-1138 (2014-09-23)
We report the isolation and complete structure of an isomer of rebaudioside D, known as rebaudioside D2. This novel steviol glycoside was isolated from a bioconversion reaction of rebaudioside A to rebaudioside D. Rebaudioside D2 possesses a relatively rare 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service