Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152110

Sigma-Aldrich

10-Chloro-9-anthraldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9ClO
CAS Number:
Molecular Weight:
240.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

215-218 °C (lit.)

functional group

aldehyde
chloro

SMILES string

[H]C(=O)c1c2ccccc2c(Cl)c3ccccc13

InChI

1S/C15H9ClO/c16-15-12-7-3-1-5-10(12)14(9-17)11-6-2-4-8-13(11)15/h1-9H

InChI key

SHYBXXMECBHHFH-UHFFFAOYSA-N

Application

10-Chloro-9-anthraldehyde was used in the preparation of trans-1-(10-methyl-9-anthryl)-2-(4-pyridyl)-ethene, 1-(10-chloro-9-anthryl)-2-(4-pyridyl)-ethene, 1-(10-methyl-9-anthryl)-2-(4-(N-methyl)-pyridinium)ethene iodide and 1-(10-chloro-9-anthryl)-2-(4-(N-methyl)pyridinium)ethene iodide[1].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substituent Effect on Fluorescence and Photoisomerization of 1-(9-Anthryl)-2-(4-pyridyl) ethenes.
Bull. Korean Chem. Soc., 23(9), 1309-1314 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service