Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15058

Sigma-Aldrich

(2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

≥98.0%

Synonym(s):

Boc-(2R,3R)-3-amino-2-hydroxy-4-phenylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO5
CAS Number:
Molecular Weight:
295.33
Beilstein/REAXYS Number:
8993437
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Product Name

(2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, ≥98.0%

assay

≥98.0%

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H](Cc1ccccc1)[C@@H](O)C(O)=O

InChI

1S/C15H21NO5/c1-15(2,3)21-14(20)16-11(12(17)13(18)19)9-10-7-5-4-6-8-10/h4-8,11-12,17H,9H2,1-3H3,(H,16,20)(H,18,19)/t11-,12-/m1/s1

InChI key

BHTRKISIDQZUQX-VXGBXAGGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service