15037
Boc-Asp-NH2
≥98.0% (TLC)
Synonym(s):
Boc-L-aspartic acid 1-amide, Boc-L-isoasparagine
About This Item
Recommended Products
assay
≥98.0% (TLC)
optical activity
[α]20/D −29±2°, c = 1% in DMF
reaction suitability
reaction type: Boc solid-phase peptide synthesis
mp
150-154 °C
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)N[C@@H](CC(O)=O)C(N)=O
InChI
1S/C9H16N2O5/c1-9(2,3)16-8(15)11-5(7(10)14)4-6(12)13/h5H,4H2,1-3H3,(H2,10,14)(H,11,15)(H,12,13)/t5-/m0/s1
InChI key
VKCARTLEXJLJBZ-YFKPBYRVSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service