About This Item
Recommended Products
assay
≥99%
form
solid
reaction suitability
core: tin
availability
available only in EU
refractive index
n20/D 1.441 (lit.)
bp
74-75 °C (lit.)
mp
−54 °C (lit.)
density
1.291 g/mL at 25 °C (lit.)
SMILES string
C[Sn](C)(C)C
InChI
1S/4CH3.Sn/h4*1H3;
Inchi Key
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
10.4 °F - closed cup
flash_point_c
-12 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service