Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

145912

Sigma-Aldrich

Quinine hemisulfate salt monohydrate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47
Beilstein/REAXYS Number:
6113937
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

90%

optical activity

[α]25/D −229°, c = 2 in 0.1 M HCl

mp

~225 °C (dec.) (lit.)
233-235 °C (dec.)

antibiotic activity spectrum

parasites

mode of action

enzyme | inhibits

SMILES string

O.O.OS(O)(=O)=O.COc1ccc2nccc([C@@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)C7CC8CCN7C[C@@H]8C=C)c6c5

InChI

1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1

InChI key

ZHNFLHYOFXQIOW-LPYZJUEESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potassium channel blocker. Antimalarial
Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive, and hypoglycemic agent; alkaloid originally isolated from the Cinchona family of South American trees. Inhibits mitochondrial ATP-regulated potassium channel. Used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Other Notes

Contains hydroquinine sulfate monohydrate

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zachary R Churcher et al.
Biophysical journal, 119(6), 1147-1156 (2020-09-04)
We have used magnetization transfer NMR experiments to measure the exchange rate constant (kex) of the imino protons in the unbound, cocaine-bound, and quinine-bound forms of the cocaine-binding DNA aptamer. Both long-stem 1 (MN4) and short-stem 1 (MN19) variants were
J P Mooney et al.
Mucosal immunology, 7(6), 1302-1311 (2014-03-29)
Coinfection can markedly alter the response to a pathogen, thereby changing its clinical presentation. For example, non-typhoidal Salmonella (NTS) serotypes are associated with gastroenteritis in immunocompetent individuals. In contrast, individuals with severe pediatric malaria can develop bacteremic infections with NTS
Eva Woltmann et al.
Analytical and bioanalytical chemistry, 406(25), 6347-6362 (2014-08-12)
High production output of solid pharmaceutical formulations requires fast methods to ensure their quality. Likewise, fast analytical procedures are required in forensic sciences, for example at customs, to substantiate an initial suspicion. We here present the design and the optimization
Jie Ma et al.
Talanta, 143, 205-211 (2015-06-17)
Hollow cross-linked fluorescent carbon nitride nanoparticles (CNNPs) were fabricated via a facile one-pot solvothermal process. The obtained CNNPs were characterized by multiple analytical techniques including transmission electron microscopy (TEM), X-ray diffraction (XRD), X-ray photoelectron spectroscopy (XPS), solid-state nuclear magnetic resonance
Joe Briscoe et al.
Angewandte Chemie (International ed. in English), 54(15), 4463-4468 (2015-02-24)
New hybrid materials consisting of ZnO nanorods sensitized with three different biomass-derived carbon quantum dots (CQDs) were synthesized, characterized, and used for the first time to build solid-state nanostructured solar cells. The performance of the devices was dependent on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service