About This Item
Recommended Products
assay
97%
form
liquid
refractive index
n20/D 1.623 (lit.)
bp
119-120 °C/10 mmHg (lit.)
density
1.407 g/mL at 25 °C (lit.)
SMILES string
Sc1ccc(Cl)c(Cl)c1
InChI
1S/C6H4Cl2S/c7-5-2-1-4(9)3-6(5)8/h1-3,9H
InChI key
HNJZDPKMMZXSKT-UHFFFAOYSA-N
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
>235.4 °F - closed cup
flash_point_c
> 113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service