144193
4,6-Dichloro-2-methylthio-5-phenylpyrimidine
95%
About This Item
Recommended Products
assay
95%
mp
107-111 °C (lit.)
functional group
chloro
phenyl
thioether
SMILES string
CSc1nc(Cl)c(c(Cl)n1)-c2ccccc2
InChI
1S/C11H8Cl2N2S/c1-16-11-14-9(12)8(10(13)15-11)7-5-3-2-4-6-7/h2-6H,1H3
Inchi Key
LMTMZMBTPXASSW-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service