Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

143251

Sigma-Aldrich

3-Amino-2,6-dimethoxypyridine monohydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2 · HCl
CAS Number:
Molecular Weight:
190.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

powder

mp

212 °C (lit.)

SMILES string

Cl[H].COc1ccc(N)c(OC)n1

InChI

1S/C7H10N2O2.ClH/c1-10-6-4-3-5(8)7(9-6)11-2;/h3-4H,8H2,1-2H3;1H

InChI key

FITCPYYVAJWASG-UHFFFAOYSA-N

Application

3-Amino-2,6-dimethoxypyridine monohydrochloride was used to investigate the potential matrices for matrix-assisted laser desorption/ionization analysis of proteins and oligonucleotides.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Fitzgerald et al.
Analytical chemistry, 65(22), 3204-3211 (1993-11-15)
In order to examine the importance of pH in the matrix-assisted laser desorption/ionization (MALDI) analysis of proteins and oligonucleotides, 37 highly substituted pyrimidine, pyridine, and benzene derivatives containing basic amino groups were screened as potential matrices. Several of these compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service