Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139025

Sigma-Aldrich

o-Arsanilic acid

98%

Synonym(s):

2-Aminobenzenearsonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4AsO3H2
CAS Number:
Molecular Weight:
217.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

mp

154-155 °C (lit.)

solubility

1 M NH4OH: soluble 50 mg/mL, clear to turbid (orange-brown)

SMILES string

Nc1ccccc1[As](O)(O)=O

InChI

1S/C6H8AsNO3/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H,8H2,(H2,9,10,11)

InChI key

LQCOCUQCZYAYQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Arsanilic acid on condensation reaction with methacryloyl chloride yields o-methacryloylaminophenylarsonic acid.

Application

o-Arsanilic acid was used in the determination of total arsenic in urine samples using nickel and palladium as matrix modifiers. It was used to study the inhibition of the binding of labeled anti-cross-reactive idiotype(CRI) preparations to a CRI+ hybridoma proteins by haptens in mice.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Nelles et al.
The Journal of experimental medicine, 154(6), 1752-1763 (1981-12-01)
Previous reports have shown the A/J anti-para-azophenylarsonate (anti-Ar) antibodies that share a major cross-reactive idiotype (CRI) comprise a family of closely related but nonidentical molecules. Serological studies with CRI+ monoclonal anti-Ar antibodies have suggested the presence of a conserved idiotypic
Crystal and molecular structure of o-methacryloylaminophenylarsonic acid.
Percino MJ, et al.
Journal of Chemical Crystallography, 32(9), 307-314 (2002)
D E Nixon et al.
Clinical chemistry, 37(9), 1575-1579 (1991-09-01)
We evaluated the effectiveness of nickel and palladium with or without added potassium persulfate as matrix modifiers for the determination of total arsenic in urine. Complete recovery of pure aqueous solutions of As(III), As(V), cacodylic acid (DMA), monomethylarsinic acid (MMA)
Han Young Eom et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1006, 151-157 (2015-11-10)
A simple and sensitive derivatization method using toluene-3,4-dithiol as a derivatization reagent for the simultaneous analysis of seven arsenic compounds (roxarsone, nitarsone, p-arsanilic acid, o-arsanilic acid, phenylarsonic acid, phenylarsine oxide, and mono-methylarsonic acid) in chicken muscle was developed and validated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service