Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

138029

Sigma-Aldrich

Furil

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6O4
CAS Number:
Molecular Weight:
190.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

163-165 °C (lit.)

SMILES string

O=C(C(=O)c1ccco1)c2ccco2

InChI

1S/C10H6O4/c11-9(7-3-1-5-13-7)10(12)8-4-2-6-14-8/h1-6H

InChI key

SXPUVBFQXJHYNS-UHFFFAOYSA-N

Gene Information

General description

Furil undergoes catalytic asymmetric transfer hydrogenation to yield hydrofuroin[1].

Application

Furil was used in preparation of dioxomolybdenum(VI) complexes in situ[2].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses and spectroscopic studies on macrocyclic complexes of dioxomolybdenum (VI) with furil as precursor.
Rao DP, et al.
E-Journal of Chemistry, 9(2), 497-503 (2012)
Anzhelika Kabro et al.
Organic letters, 14(15), 4014-4017 (2012-07-27)
Catalytic asymmetric transfer hydrogenation of rac-furoin and furil produces hydrofuroin with up to 99% ee and 9:1 dr. This reaction provides an exceptionally easy access to optically active hydrofuroins in two straightforward steps from biomass-derived furfural (global production 200,000-300,000 t

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service