About This Item
Recommended Products
assay
97%
form
liquid
refractive index
n20/D 1.52 (lit.)
bp
195 °C (lit.)
mp
29 °C (lit.)
density
0.984 g/mL at 25 °C (lit.)
SMILES string
COc1cccc(C)c1C
InChI
1S/C9H12O/c1-7-5-4-6-9(10-3)8(7)2/h4-6H,1-3H3
Inchi Key
BLMBNEVGYRXFNA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service